siRNA Details
| Gene Symbol: PDCD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199492.1 | NP_001186421.1 | 12 | 1990 | ||
| NM_014456.4 | NP_055271.2 | 12 | 2032 | |||
| NM_145341.3 | NP_663314.1 | 13 | 2116 | |||
| GenBank mRNA | AK223623.1 | - | 12 | 1910 | ||
| AL049932.1 | - | 1 | 265 | |||
| BC026104.2 | - | 12 | 1898 | 4798293 | ||
| BC031049.1 | - | 12 | 1897 | 5277621 | ||
| BX537500.1 | - | 10 | 5904 | |||
| BX647834.1 | - | 12 | 1872 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001168491, NM_011050, D50465, AK050175, D86344, and others.

PDCD4
Genomic Map