siRNA Details
Gene Symbol: Slc22a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138908.2 | NP_620263.1 | 2, 3 | 598 | ||
GenBank mRNA | Y09945.1 | - | 2, 3 | 600 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138908.2 | NP_620263.1 | 2, 3 | 598 | ||
GenBank mRNA | Y09945.1 | - | 2, 3 | 600 |