siRNA Details
| Gene Symbol: Unc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138919.2 | NP_620274.1 | 2 | 352 | ||
| XM_006244736.2 | XP_006244798.1 | 2 | 305 | |||
| XM_006244737.2 | XP_006244799.1 | 2 | 250 | |||
| GenBank mRNA | AY017209.1 | - | 2 | 352 | ||
| BC061976.1 | - | 2 | 347 | 5623807 | ||
| FQ220352.1 | - | 2 | 246 | |||
| U96638.1 | - | 1 | 118 | |||

Unc50
Genomic Map