siRNA Details
| Gene Symbol: Rabggtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138708.2 | NP_619715.1 | 5 | 518 | ||
| GenBank mRNA | BC060536.1 | - | 542 | |||
| BC078683.1 | - | 5 | 507 | 7135106 | ||
| FQ214773.1 | - | 522 | ||||
| FQ216744.1 | - | 522 | ||||
| FQ224712.1 | - | 521 | ||||
| FQ229109.1 | - | 522 | ||||
| L10416.1 | - | 5 | 518 | |||
| S62097.1 | - | 5 | 518 | |||

Rabggtb
Genomic Map