siRNA Details
| Gene Symbol: Ubxn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138853.2 | NP_620208.1 | 4, 5 | 267 | ||
| XM_006239000.1 | XP_006239062.1 | 4, 5 | 329 | |||
| XM_006239002.1 | XP_006239064.1 | 4, 5 | 329 | |||
| XM_006239004.1 | XP_006239066.1 | 2, 3 | 183 | |||
| XM_008764131.1 | XP_008762353.1 | 2, 3 | 335 | |||
| XM_008764132.1 | XP_008762354.1 | 2, 3 | 376 | |||
| GenBank mRNA | AB072920.1 | - | 3, 4 | 195 | ||
| BC078730.1 | - | 4, 5 | 267 | 7134156 | ||

Ubxn11
Genomic Map