siRNA Details
| Gene Symbol: Akr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138884.1 | NP_620239.1 | 2 | 258 | ||
| XM_006236285.1 | XP_006236347.1 | 2 | 278 | |||
| GenBank mRNA | D17309.1 | - | 2 | 258 | ||
| FQ211413.1 | - | 2 | 276 | |||
| FQ219185.1 | - | 2 | 277 | |||
| FQ219421.1 | - | 2 | 221 | |||
| FQ219730.1 | - | 2 | 221 | |||
| S80431.1 | - | 2 | 230 | |||

Akr1d1
Genomic Map