siRNA Details
| Gene Symbol: Stip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138911.3 | NP_620266.1 | 2, 3 | 292 | ||
| GenBank mRNA | BC061529.1 | - | 2, 3 | 293 | 6887497 | |
| Y15068.1 | - | 1, 2 | 227 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stip1| Gene Symbol: Stip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138911.3 | NP_620266.1 | 2, 3 | 292 | ||
| GenBank mRNA | BC061529.1 | - | 2, 3 | 293 | 6887497 | |
| Y15068.1 | - | 1, 2 | 227 | |||