siRNA Details
| Gene Symbol: Fat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138544.1 | NP_612553.1 | 11 | 9027 | ||
| XM_008765910.1 | XP_008764132.1 | 9 | 7848 | |||
| XM_008765911.1 | XP_008764133.1 | 9 | 7848 | |||
| XM_008765912.1 | XP_008764134.1 | 9 | 7848 | |||
| XM_008765913.1 | XP_008764135.1 | 9 | 7848 | |||
| XM_008765914.1 | XP_008764136.1 | 8 | 7779 | |||
| XM_008765915.1 | XP_008764137.1 | 9 | 7848 | |||
| XM_008765916.1 | XP_008764138.1 | 9 | 7848 | |||
| GenBank mRNA | AB076401.1 | - | 11 | 9027 | ||

Fat3
Genomic Map