siRNA Details
| Gene Symbol: Dnajc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138856.2 | NP_620211.2 | 7 | 709 | ||
| XM_008760746.1 | XP_008758968.1 | 11 | 1585 | |||
| GenBank mRNA | AF492384.1 | - | 6 | 632 | ||
| BC086587.1 | - | 7 | 709 | |||
| BC158554.1 | - | 7 | 928 | 7380100 | ||

Dnajc21
Genomic Map