siRNA Details
Gene Symbol: Prl2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138861.1 | NP_620216.1 | 5 | 703 | ||
GenBank mRNA | AB022882.1 | - | 5 | 700 | ||
AF234635.1 | - | 5 | 703 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prl2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138861.1 | NP_620216.1 | 5 | 703 | ||
GenBank mRNA | AB022882.1 | - | 5 | 700 | ||
AF234635.1 | - | 5 | 703 |