siRNA Details
| Gene Symbol: Pigl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138901.2 | NP_620256.1 | 5, 6 | 543 | ||
| XM_008767767.1 | XP_008765989.1 | 5, 6 | 539 | |||
| GenBank mRNA | BC074020.1 | - | 5, 6 | 543 | 7104008 | |
| D88364.1 | - | 5, 6 | 924 | |||

Pigl
Genomic Map