siRNA Details
| Gene Symbol: BRINP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014618.2 | NP_055433.2 | 8 | 2872 | ||
| GenBank mRNA | AF027734.1 | - | 8 | 2834 | ||
| AK293649.1 | - | 4 | 1922 | |||
| BC065196.1 | - | 8 | 2825 | 5245480 | ||
| BC071702.1 | - | 4 | 1820 | 5244680 | ||

BRINP1
Genomic Map