siRNA Details
| Gene Symbol: Ss18l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138918.1 | NP_620273.1 | 7 | 783 | ||
| GenBank mRNA | AY034073.1 | - | 7 | 783 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ss18l1| Gene Symbol: Ss18l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138918.1 | NP_620273.1 | 7 | 783 | ||
| GenBank mRNA | AY034073.1 | - | 7 | 783 | ||