siRNA Details
Gene Symbol: Prokr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138977.1 | NP_620433.1 | 1 | 155 | ||
XM_006236821.2 | XP_006236883.1 | 2 | 450 | |||
GenBank mRNA | AB158419.1 | - | 1 | 155 | ||
AB158420.1 | - | 1 | 155 | |||
AY089974.1 | - | 1 | 155 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
