siRNA Details
| Gene Symbol: Impg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139088.2 | NP_620788.2 | 2 | 323 | ||
| XM_008768628.1 | XP_008766850.1 | 2 | 1311 | |||
| GenBank mRNA | U76717.1 | - | 2 | 323 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Impg2| Gene Symbol: Impg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139088.2 | NP_620788.2 | 2 | 323 | ||
| XM_008768628.1 | XP_008766850.1 | 2 | 1311 | |||
| GenBank mRNA | U76717.1 | - | 2 | 323 | ||