siRNA Details
| Gene Symbol: Gpr116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139110.1 | NP_620810.1 | 3, 4 | 329 | ||
| XM_006244594.2 | XP_006244656.1 | 3, 4 | 544 | |||
| XM_006244597.2 | XP_006244659.1 | 3, 4 | 543 | |||
| XM_008766832.1 | XP_008765054.1 | 3, 4 | 546 | |||
| XM_008766833.1 | XP_008765055.1 | 3, 4 | 471 | |||
| XM_008766834.1 | XP_008765056.1 | 3, 4 | 547 | |||
| XM_008766835.1 | XP_008765057.1 | 3, 4 | 545 | |||
| GenBank mRNA | AB019120.1 | - | 3, 4 | 329 | ||

Gpr116
Genomic Map