siRNA Details
| Gene Symbol: Crhbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139183.2 | NP_631922.1 | 3 | 396 | ||
| GenBank mRNA | X58023.1 | - | 3 | 395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crhbp| Gene Symbol: Crhbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139183.2 | NP_631922.1 | 3 | 396 | ||
| GenBank mRNA | X58023.1 | - | 3 | 395 | ||