siRNA Details
| Gene Symbol: Mcfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139253.1 | NP_640346.1 | 3 | 256 | ||
| XM_006239710.2 | XP_006239772.1 | 3 | 295 | |||
| XM_006239711.2 | XP_006239773.1 | 3 | 316 | |||
| GenBank mRNA | AF475282.1 | - | 3 | 256 | ||
| BC071179.1 | - | 3 | 250 | 7100216 | ||

Mcfd2
Genomic Map