siRNA Details
| Gene Symbol: Bmf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139258.1 | NP_640351.1 | 4 | 616 | ||
| XM_006234723.2 | XP_006234785.1 | 4 | 654 | |||
| XM_006234724.2 | XP_006234786.1 | 4 | 657 | |||
| GenBank mRNA | AF506761.1 | - | 4 | 616 | ||

Bmf
Genomic Map