siRNA Details
| Gene Symbol: Slc34a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139338.2 | NP_647554.1 | 5 | 418 | ||
| XM_006233558.2 | XP_006233620.1 | 6 | 631 | |||
| XM_006233559.2 | XP_006233621.1 | 6 | 747 | |||
| XM_006233560.2 | XP_006233622.1 | 6 | 608 | |||
| XM_006233561.2 | XP_006233623.1 | 5 | 521 | |||
| XM_006233562.2 | XP_006233624.1 | 5 | 637 | |||
| XM_006233564.2 | XP_006233626.1 | 5 | 501 | |||
| XM_006233565.2 | XP_006233627.1 | 6 | 607 | |||
| XM_006233566.2 | XP_006233628.1 | 5 | 613 | |||
| XM_008761577.1 | XP_008759799.1 | 6 | 696 | |||
| GenBank mRNA | AB077042.1 | - | 5 | 423 | ||

Slc34a3
Genomic Map