siRNA Details
Gene Symbol: Adipoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144744.3 | NP_653345.1 | 2 | 192 | ||
GenBank mRNA | AY033885.1 | - | 2 | 139 | ||
BC092565.1 | - | 1 | 128 | 7308618 | ||
FM047633.1 | - | 2 | 192 |
