siRNA Details
| Gene Symbol: Gpsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144745.2 | NP_653346.2 | 2 | 261 | ||
| XM_006233670.2 | XP_006233732.1 | 2 | 365 | |||
| GenBank mRNA | AF107723.1 | - | 1 | 385 | ||
| CB743977.1 | - | 2 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpsm1| Gene Symbol: Gpsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144745.2 | NP_653346.2 | 2 | 261 | ||
| XM_006233670.2 | XP_006233732.1 | 2 | 365 | |||
| GenBank mRNA | AF107723.1 | - | 1 | 385 | ||
| CB743977.1 | - | 2 | 309 | |||