siRNA Details
| Gene Symbol: RGD621098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139040.1 | NP_620609.1 | 2 | 347 | ||
| GenBank mRNA | U92010.1 | - | 2 | 347 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD621098| Gene Symbol: RGD621098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139040.1 | NP_620609.1 | 2 | 347 | ||
| GenBank mRNA | U92010.1 | - | 2 | 347 | ||