siRNA Details
| Gene Symbol: Rnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270762.1 | NP_001257691.1 | 5 | 696 | ||
| NM_001270763.1 | NP_001257692.1 | 5 | 702 | |||
| NM_139105.3 | NP_620805.2 | 5 | 711 | |||
| XM_008759970.1 | XP_008758192.1 | 5 | 1260 | |||
| GenBank mRNA | AB296102.1 | - | 4 | 578 | ||
| BC070501.1 | - | 5 | 618 | 7096144 | ||
| FQ233708.1 | - | 5 | 636 | |||
| FQ234889.1 | - | 5 | 635 | |||
| X62528.1 | - | 5 | 696 | |||

Rnh1
Genomic Map