siRNA Details
| Gene Symbol: Nradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139259.2 | NP_640352.1 | 4 | 497 | ||
| XM_006243921.2 | XP_006243983.1 | 4 | 501 | |||
| GenBank mRNA | AF497263.1 | - | 4 | 501 | ||
| AF497264.1 | - | 4 | 349 | |||
| AF534395.1 | - | 3 | 313 | |||

Nradd
Genomic Map