siRNA Details
| Gene Symbol: Prpf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139333.3 | NP_647549.2 | 7 | 732 | ||
| XM_006231013.2 | XP_006231075.1 | 7 | 825 | |||
| XM_006231014.2 | XP_006231076.1 | 7 | 823 | |||
| XM_006231015.2 | XP_006231077.1 | 7 | 767 | |||
| GenBank mRNA | AB020022.2 | - | 7 | 778 | ||
| BC107669.1 | - | 7 | 732 | 7937645 | ||

Prpf19
Genomic Map