siRNA Details
| Gene Symbol: Pnpla7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144738.2 | NP_653339.2 | 20, 21 | 2411 | ||
| GenBank mRNA | AY100477.1 | - | 2, 3 | 213 | ||
| BC083547.1 | - | 3, 4 | 217 | 7132749 | ||
| BC091230.1 | - | 20, 21 | 2411 | 7321805 | ||

Pnpla7
Genomic Map