siRNA Details
| Gene Symbol: Adipoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144744.3 | NP_653345.1 | 2, 3 | 297 | ||
| GenBank mRNA | AY033885.1 | - | 2, 3 | 244 | ||
| BC092565.1 | - | 1, 2 | 233 | 7308618 | ||
| FM047633.1 | - | 2, 3 | 297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adipoq| Gene Symbol: Adipoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144744.3 | NP_653345.1 | 2, 3 | 297 | ||
| GenBank mRNA | AY033885.1 | - | 2, 3 | 244 | ||
| BC092565.1 | - | 1, 2 | 233 | 7308618 | ||
| FM047633.1 | - | 2, 3 | 297 | |||