siRNA Details
Gene Symbol: Lhx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139036.1 | NP_620605.1 | 5 | 1239 | ||
GenBank mRNA | L35572.1 | - | 5 | 1239 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lhx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139036.1 | NP_620605.1 | 5 | 1239 | ||
GenBank mRNA | L35572.1 | - | 5 | 1239 |