siRNA Details
Gene Symbol: Muc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768788.1 | XP_008767010.1 | 10, 11 | 1510 | ||
GenBank mRNA | U89744.1 | - | 9, 10 | 1472 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
