siRNA Details
| Gene Symbol: Exoc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139043.1 | NP_620612.1 | 1 | 1813 | ||
| GenBank mRNA | AF032669.1 | - | 1 | 1813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exoc8| Gene Symbol: Exoc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139043.1 | NP_620612.1 | 1 | 1813 | ||
| GenBank mRNA | AF032669.1 | - | 1 | 1813 | ||