siRNA Details
| Gene Symbol: Xrcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139080.2 | NP_620780.2 | 11 | 1616 | ||
| XM_008765671.1 | XP_008763893.1 | 10 | 1447 | |||
| GenBank mRNA | AB066102.1 | - | 10 | 1600 | ||
| BC078718.1 | - | 11 | 1616 | 7115290 | ||

Xrcc6
Genomic Map