siRNA Details
| Gene Symbol: FEM1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015322.4 | NP_056137.1 | 2 | 1772 | ||
| XM_017021859.1 | XP_016877348.1 | 2 | 1285 | |||
| GenBank mRNA | AB007856.2 | - | 2 | 1157 | ||
| AF178632.1 | - | 2 | 1157 | |||
| AF204883.1 | - | 2 | 1157 | |||
| AK098620.1 | - | 1 | 293 | |||
| AK290167.1 | - | 2 | 1506 | |||
| AL833264.1 | - | 1 | 2104 | |||
| BC010122.2 | - | 2 | 1772 | 3840453 | ||
| BC014558.2 | - | 2 | 1772 | 3840377 | ||

FEM1B
Genomic Map