siRNA Details
| Gene Symbol: Impg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139088.2 | NP_620788.2 | 14 | 2687 | ||
| XM_008768628.1 | XP_008766850.1 | 14 | 3675 | |||
| XM_008768629.1 | XP_008766851.1 | 9 | 2035 | |||
| GenBank mRNA | U76717.1 | - | 14 | 2687 | ||

Impg2
Genomic Map