siRNA Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139091.2 | NP_620791.1 | 8 | 840 | ||
| XM_006252093.2 | XP_006252155.1 | 8 | 945 | |||
| GenBank mRNA | AF000900.1 | - | 7 | 711 | ||
| AF000901.1 | - | 7 | 733 | |||
| BC085690.1 | - | 8 | 840 | 7190650 | ||
| U63839.1 | - | 8 | 853 | |||

Nupl1
Genomic Map