siRNA Details
| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139109.2 | NP_620809.1 | 6 | 756 | ||
| GenBank mRNA | AF277899.1 | - | 6 | 759 | ||
| BC081723.1 | - | 6 | 750 | 7108549 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xab2| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139109.2 | NP_620809.1 | 6 | 756 | ||
| GenBank mRNA | AF277899.1 | - | 6 | 759 | ||
| BC081723.1 | - | 6 | 750 | 7108549 | ||