siRNA Details
| Gene Symbol: Ppap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139252.1 | NP_640345.1 | 6 | 864 | ||
| XM_008765155.1 | XP_008763377.1 | 6 | 847 | |||
| GenBank mRNA | AF503611.1 | - | 6 | 864 | ||
| BC062088.1 | - | 6 | 820 | 5621318 | ||

Ppap2c
Genomic Map