siRNA Details
| Gene Symbol: Hspbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139261.1 | NP_640354.1 | 6 | 970 | ||
| XM_006228248.1 | XP_006228310.1 | 6 | 1197 | |||
| XM_006228249.2 | XP_006228311.1 | 6 | 962 | |||
| XM_006228250.2 | XP_006228312.1 | 6 | 1026 | |||
| GenBank mRNA | AF187860.1 | - | 6 | 970 | ||
| BC072541.1 | - | 6 | 1006 | 7103744 | ||
| FQ232199.1 | - | 6 | 1026 | |||

Hspbp1
Genomic Map