siRNA Details
| Gene Symbol: Fgd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139263.1 | NP_640356.1 | 16 | 3207 | ||
| XM_008768850.1 | XP_008767072.1 | 15 | 2511 | |||
| GenBank mRNA | AF038388.1 | - | 16 | 3207 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgd4| Gene Symbol: Fgd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139263.1 | NP_640356.1 | 16 | 3207 | ||
| XM_008768850.1 | XP_008767072.1 | 15 | 2511 | |||
| GenBank mRNA | AF038388.1 | - | 16 | 3207 | ||