siRNA Details
| Gene Symbol: Tas2r126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139335.1 | NP_647551.1 | 1 | 490 | ||
| GenBank mRNA | AF240768.1 | - | 1 | 490 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r126| Gene Symbol: Tas2r126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139335.1 | NP_647551.1 | 1 | 490 | ||
| GenBank mRNA | AF240768.1 | - | 1 | 490 | ||