siRNA Details
| Gene Symbol: Ripk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139342.1 | NP_647558.1 | 9, 10 | 1367 | ||
| XM_006251928.2 | XP_006251990.1 | 8, 9 | 2219 | |||
| XM_008770661.1 | XP_008768883.1 | 7, 8 | 1180 | |||
| XM_008770662.1 | XP_008768884.1 | 8, 9 | 1551 | |||
| GenBank mRNA | AF036537.2 | - | 9, 10 | 1367 | ||
| BC158580.1 | - | 7, 8 | 934 | 7378607 | ||

Ripk3
Genomic Map