siRNA Details
| Gene Symbol: LOC246267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144751.1 | NP_653352.1 | 92 | |||
| GenBank mRNA | AF007890.1 | - | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC246267| Gene Symbol: LOC246267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144751.1 | NP_653352.1 | 92 | |||
| GenBank mRNA | AF007890.1 | - | 92 | |||