siRNA Details
| Gene Symbol: Cant1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144754.2 | NP_653355.1 | 2 | 332 | ||
| XM_006247789.2 | XP_006247851.1 | 2 | 854 | |||
| XM_006247790.1 | XP_006247852.1 | 2 | 402 | |||
| GenBank mRNA | AJ312207.1 | - | 1 | 287 | ||
| BC097279.1 | - | 2 | 332 | 7443057 | ||

Cant1
Genomic Map