siRNA Details
| Gene Symbol: Asrgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145089.4 | NP_659557.1 | 2 | 222 | ||
| GenBank mRNA | AF329099.1 | - | 2 | 222 | ||
| AJ427914.2 | - | 236 | ||||
| FQ212762.1 | - | 2 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asrgl1| Gene Symbol: Asrgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145089.4 | NP_659557.1 | 2 | 222 | ||
| GenBank mRNA | AF329099.1 | - | 2 | 222 | ||
| AJ427914.2 | - | 236 | ||||
| FQ212762.1 | - | 2 | 196 | |||