siRNA Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762466.1 | XP_008760688.1 | 5, 6 | 938 | ||
| GenBank mRNA | AB029897.1 | - | 3, 4 | 446 | ||
| AB180269.1 | - | 3, 4 | 446 | |||
| AB180270.1 | - | 3, 4 | 446 | |||
| AB180271.1 | - | 3, 4 | 446 | |||
| BC094306.1 | - | 3, 4 | 434 | |||
| CO405877.1 | - | 1, 2 | 32 | |||

Bcas1
Genomic Map