siRNA Details
| Gene Symbol: Cxcl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145672.4 | NP_663705.1 | 2 | 179 | ||
| GenBank mRNA | AF462610.1 | - | 2 | 86 | ||
| AF537208.1 | - | 2 | 179 | |||
| BC087594.1 | - | 128 | 7310193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl9| Gene Symbol: Cxcl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145672.4 | NP_663705.1 | 2 | 179 | ||
| GenBank mRNA | AF462610.1 | - | 2 | 86 | ||
| AF537208.1 | - | 2 | 179 | |||
| BC087594.1 | - | 128 | 7310193 | |||