siRNA Details
| Gene Symbol: Fxyd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017349.2 | NP_059045.2 | 4 | 182 | ||
| NM_145717.1 | NP_663769.1 | 5 | 226 | |||
| XM_006242911.2 | XP_006242973.1 | 5 | 383 | |||
| XM_006242912.2 | XP_006242974.1 | 5 | 331 | |||
| GenBank mRNA | AF129400.1 | - | 5 | 226 | ||
| AF233060.1 | - | 4 | 182 | |||
| X70062.1 | - | 5 | 190 | |||

Fxyd2
Genomic Map