siRNA Details
| Gene Symbol: Tnfsf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145765.1 | NP_665708.1 | 4 | 444 | ||
| GenBank mRNA | AF520787.1 | - | 4 | 444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfsf15| Gene Symbol: Tnfsf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145765.1 | NP_665708.1 | 4 | 444 | ||
| GenBank mRNA | AF520787.1 | - | 4 | 444 | ||