siRNA Details
| Gene Symbol: Lgi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145769.3 | NP_665712.1 | 2 | 261 | ||
| GenBank mRNA | AJ487517.1 | - | 2 | 261 | ||
| BC089222.1 | - | 2 | 368 | 7320899 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgi1| Gene Symbol: Lgi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145769.3 | NP_665712.1 | 2 | 261 | ||
| GenBank mRNA | AJ487517.1 | - | 2 | 261 | ||
| BC089222.1 | - | 2 | 368 | 7320899 | ||