siRNA Details
| Gene Symbol: Cox5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145783.1 | NP_665726.1 | 2, 3 | 225 | ||
| GenBank mRNA | FQ210272.1 | - | 2, 3 | 258 | ||
| FQ211124.1 | - | 2, 3 | 285 | |||
| FQ217304.1 | - | 2, 3 | 258 | |||
| FQ217698.1 | - | 2, 3 | 248 | |||
| FQ223560.1 | - | 2, 3 | 258 | |||
| FQ223883.1 | - | 2, 3 | 242 | |||
| FQ224560.1 | - | 2, 3 | 250 | |||
| FQ224768.1 | - | 2, 3 | 251 | |||
| FQ228531.1 | - | 2, 3 | 258 | |||
| X15030.1 | - | 2, 3 | 225 | |||

Cox5a
Genomic Map